missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Iodoethanol, 99%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 175500500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Iodoethanol | |
2.2050g/mL | |
65°C | |
98.5% min. (GC) | |
C2H5IO | |
ICH2CH2OH | |
50 mL | |
13,348 | |
ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol | |
OCCI | |
171.97 | |
12225 | |
99% |
624-76-0 | |
172.0°C to 175.0°C | |
Authentic | |
Glass bottle | |
1.5690 to 1.5720 | |
MFCD00002830 | |
01,339 | |
2.205 | |
QSECPQCFCWVBKM-UHFFFAOYSA-N | |
2-iodoethanol | |
Copper | |
171.97 | |
Liquid |
Chemical Identifiers
624-76-0 | |
171.97 | |
QSECPQCFCWVBKM-UHFFFAOYSA-N | |
12225 |
C2H5IO | |
MFCD00002830 | |
ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol | |
OCCI |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.
Rinse mouth.
GHS Signal Word: Danger
EINECSNumber : 210-861-6
RUO – Research Use Only