Learn More
2-Iodoethanol, 99%, stab. with copper, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1465814
Description
2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.
Solubility
Soluble in water.
Notes
Light sensitive. Store in a cool place. Incompatible with acid anhydrides, acid chlorides, phosphorus halides, strong acids, strong oxidizing agents and strong reducing agents.
Specifications
2-Iodoethanol | |
624-76-0 | |
85°C (25 mmHg) | |
Characteristic | |
1.572 | |
MFCD00002830 | |
UN2810 | |
Light sensitive | |
Soluble in water. | |
OCCI | |
171.97 | |
171.96 |
stabilized with copper | |
2.202 | |
65°C (149°F) | |
C2H5IO | |
ICH2CH2OH | |
25 g | |
1098258 | |
ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol | |
QSECPQCFCWVBKM-UHFFFAOYSA-N | |
2-iodoethanol | |
12225 | |
99% |
Chemical Identifiers
624-76-0 | |
171.97 | |
QSECPQCFCWVBKM-UHFFFAOYSA-N | |
12225 | |
OCCI |
C2H5IO | |
MFCD00002830 | |
ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol | |
2-iodoethanol |
Safety and Handling
GHS H Statement
H300-H310-H341-H227
Fatal if swallowed.
Fatal in contact with skin.
Suspected of causing genetic defects.
Combustible liquid.
P210-P264b-P270-P280-P301+P310-P330-P370+P378q-P501c
H227-H300
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 210-861-6
RTECSNumber : KL3820000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only