missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iododibenzothiophene, TCI America™

$99.31 - $342.23

Chemical Identifiers

CAS 177586-41-3
Molecular Formula C12H7IS
Molecular Weight (g/mol) 310.152
InChI Key LCTVJYHYWSCJAF-UHFFFAOYSA-N
PubChem CID 21709598
IUPAC Name 2-iododibenzothiophene
SMILES C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0973200MG View Documents TCI America
I0973200MG
200mg Each for $99.31
 
I09731G View Documents TCI America
I09731G
1g Each for $342.23
 
Specifications

Chemical Identifiers

177586-41-3
310.152
21709598
C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)I
C12H7IS
LCTVJYHYWSCJAF-UHFFFAOYSA-N
2-iododibenzothiophene

Specifications

White-Yellow
200mg
C12H7IS
C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)I
310.152
310.15
≥98.0% (GC)
88°C
177586-41-3
LCTVJYHYWSCJAF-UHFFFAOYSA-N
2-iododibenzothiophene
21709598
Crystal-Powder at 20°C
2-Iododibenzothiophene
Safety and Handling

Safety and Handling

TSCA : No