missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodocumene, TCI America™

$51.95 - $151.25

Chemical Identifiers

CAS 19099-54-8
Molecular Formula C9H11I
Molecular Weight (g/mol) 246.091
MDL Number MFCD00013704
InChI Key SORQIYFSJAWBNQ-UHFFFAOYSA-N
Synonym 1-iodo-2-isopropylbenzene, 2-iodoisopropylbenzene, 2-iodocumene, o-iodocumene, 1-iodo-2-isopropyl-benzene, benzene, 1-iodo-2-1-methylethyl, o-i-propyliodobenzene, 2-isopropyliodobenzene, 2-iodoisopropyl benzene, pubchem12584
PubChem CID 87931
IUPAC Name 1-iodo-2-propan-2-ylbenzene
SMILES CC(C)C1=CC=CC=C1I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I05595G View Documents TCI America
I05595G
5g Each for $51.95
 
I055925G View Documents TCI America
I055925G
25g Each for $151.25
 
Specifications

Chemical Identifiers

19099-54-8
246.091
SORQIYFSJAWBNQ-UHFFFAOYSA-N
87931
CC(C)C1=CC=CC=C1I
C9H11I
MFCD00013704
1-iodo-2-isopropylbenzene, 2-iodoisopropylbenzene, 2-iodocumene, o-iodocumene, 1-iodo-2-isopropyl-benzene, benzene, 1-iodo-2-1-methylethyl, o-i-propyliodobenzene, 2-isopropyliodobenzene, 2-iodoisopropyl benzene, pubchem12584
1-iodo-2-propan-2-ylbenzene

Specifications

117°C
5g
C9H11I
1-iodo-2-isopropylbenzene, 2-iodoisopropylbenzene, 2-iodocumene, o-iodocumene, 1-iodo-2-isopropyl-benzene, benzene, 1-iodo-2-1-methylethyl, o-i-propyliodobenzene, 2-isopropyliodobenzene, 2-iodoisopropyl benzene, pubchem12584
CC(C)C1=CC=CC=C1I
246.091
246.09
≥98.0% (GC)
Undesignated
19099-54-8
MFCD00013704
SORQIYFSJAWBNQ-UHFFFAOYSA-N
1-iodo-2-propan-2-ylbenzene
87931
Clear Liquid at 20°C
2-Iodocumene
Safety and Handling

Safety and Handling

TSCA : No