missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodobenzyl bromide, 97%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C7H6BrI
Formula Weight 296.93
Chemical Name or Material 2-Iodobenzyl bromide
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC443870050 SDS Thermo Fisher Scientific
AC443870050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC443870010 SDS Thermo Fisher Scientific
AC443870010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
296.93
≥96.0%
C7H6BrI
2-Iodobenzyl bromide

Specifications

40400-13-3
Crystalline Powder
C7H6BrI
White to cream
5g
≥96.0%
Authentic
97%
296.93
55° to 59°C
2-Iodobenzyl bromide

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger