Please login to your online account to display your discounted pricing

2-Iodobenzyl bromide, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC443870050

 View more versions of this product

Catalog No. AC443870050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 296.93
Quantity 5g
Color White to cream
Melting Point 55° to 59°C
Infrared Spectrum Authentic
Chemical Name or Material 2-Iodobenzyl bromide
Assay Percent Range 97%
Molecular Formula C7H6BrI
CAS 40400-13-3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Iodobenzyl bromide
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H6BrI
Formula Weight 296.93
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.