Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Iodobenzyl bromide, 96%

$107.59 - $391.76

Chemical Identifiers

Chemical Name or Material 2-Iodobenzyl bromide
CAS 40400-13-3
Molecular Formula C7H6BrI
Formula Weight 296.94
MDL Number MFCD00236046
UN Number UN3261
Beilstein 2079736
Synonym alpha-Bromo-2-iodotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2171506

  SDS
alfa aesar™
B2171506*
5g Each for $107.59

AAB2171514

  SDS
alfa aesar™
B2171514*
25g Each for $391.76

AAB2171522

  SDS
alfa aesar™
B2171522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 296.94
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H6BrI
Quantity 5g
CAS 40400-13-3
Melting Point 53° to 58°C
UN Number UN3261
Synonym alpha-Bromo-2-iodotoluene
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Assay 96%
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
MDL Number MFCD00236046
Health Hazard 1 Danger
Beilstein 2079736
Chemical Name or Material 2-Iodobenzyl bromide
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.