Learn More
2-Iodobenzoyl chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1355818
Description
2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.
Solubility
Reacts with water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
2-Iodobenzoyl chloride | |
27°C to 32°C | |
>110°C (230°F) | |
MFCD00001040 | |
UN3261 | |
Moisture sensitive | |
Reacts with water. | |
C1=CC=C(C(=C1)C(=O)Cl)I | |
266.462 | |
266.47 |
609-67-6 | |
105°C to 106°C (1 mmHg) | |
C7H4ClIO | |
50 g | |
2042672 | |
o-iodobenzoyl chloride, benzoyl chloride, 2-iodo, iodobenzoyl chloride, o-iodobenzoylchloride, 2-iodobezoyl chloride, o-iodo-benzoylchloride, 2-iodobenzoic chloride, 2-iodo-benzoyl chloride, acmc-1atjg, benzoyl chloride,2-iodo | |
MVIVDSWUOGNODP-UHFFFAOYSA-N | |
2-iodobenzoyl chloride | |
69112 | |
98% |
Chemical Identifiers
609-67-6 | |
266.462 | |
MVIVDSWUOGNODP-UHFFFAOYSA-N | |
2-iodobenzoyl chloride |
C7H4ClIO | |
MFCD00001040 | |
69112 | |
C1=CC=C(C(=C1)C(=O)Cl)I |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 210-196-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only