Please login to your online account to display your discounted pricing

2-Iodobenzotrifluoride, 99%, ACROS Organics™

$37.45 - $37.45

Chemical Identifiers

Chemical Name or Material 2-Iodobenzotrifluoride
CAS 444-29-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H4F3I
Linear Formula CF3(C6H4)I
Formula Weight 272.01
MDL Number MFCD00001041
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC241981000

  SDS
thermo fisher scientific
AC241981000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC241980050

  SDS
thermo fisher scientific
241980050
Glass bottle 5mL Each for $37.45

AC241980250

  SDS
thermo fisher scientific
241980250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 272.01
Quantity 100mL
Color Clear light yellow
Density 1.8900g/mL
Beilstein 05, III, 725
CAS Max % 100.0
Chemical Name or Material 2-Iodobenzotrifluoride
Infrared Spectrum Authentic
Molecular Formula C7H4F3I
Linear Formula CF3(C6H4)I
CAS 444-29-1
Assay Percent Range 99%
MDL Number MFCD00001041
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.5290 to 1.5310 (20°C, 589nm)
Synonym 2-Iodo-α, α, α-trifluorotoluene
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.