missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodobenzotrifluoride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 241980050

 View more versions of this product

Catalog No. AC241980050



Specifications

Specifications

2-Iodobenzotrifluoride
81°C
Glass bottle
1.89
Yellow
444-29-1
100.0
C7H4F3I
MFCD00001041
1-iodo-2-trifluoromethyl benzene, 2-iodo-a,a,a-trifluorotoluene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, 2-iodobenzotrifloride, 2-iodobenzotrifluoride, 2-iodobenztrifluoride, benzene, 1-iodo-2-trifluoromethyl, iodobenzotrifluoride, o-iodobenzotrifluoride, o-trifluoromethyliodobenzene
IGZGUYVVBABKOY-UHFFFAOYSA-N
1-iodo-2-(trifluoromethyl)benzene
67957
Liquid
1.8900g/mL
Authentic
1.5290 to 1.5310
197.0°C to 198.0°C
5mL
98.5
98.5% min. (GC)
CF3(C6H4)I
05, III, 725
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
C1=CC=C(C(=C1)C(F)(F)F)I
272.01
272.01
99%

Chemical Identifiers

444-29-1
272.01
IGZGUYVVBABKOY-UHFFFAOYSA-N
67957
C1=CC=C(C(=C1)C(F)(F)F)I
C7H4F3I
MFCD00001041
1-iodo-2-trifluoromethyl benzene, 2-iodo-a,a,a-trifluorotoluene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, 2-iodobenzotrifloride, 2-iodobenzotrifluoride, 2-iodobenztrifluoride, benzene, 1-iodo-2-trifluoromethyl, iodobenzotrifluoride, o-iodobenzotrifluoride, o-trifluoromethyliodobenzene
1-iodo-2-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-147-1

SDS Documents