Learn More
Alfa Aesar™ 2-Iodobenzotrifluoride, 99%
Manufacturer: Alfa Aesar™ A1569822
Specifications
2-Iodobenzotrifluoride | |
198°C to 199°C | |
444-29-1 | |
MFCD00001041 | |
2-iodobenzotrifluoride, 1-iodo-2-trifluoromethyl benzene, o-iodobenzotrifluoride, benzene, 1-iodo-2-trifluoromethyl, 2-iodobenztrifluoride, o-trifluoromethyliodobenzene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, iodobenzotrifluoride, 2-iodo-a,a,a-trifluorotoluene, 2-iodobenzotrifloride | |
C1=CC=C(C(=C1)C(F)(F)F)I | |
272.009 | |
272.01 |
81°C (177°F) | |
100g | |
C7H4F3I | |
2090038 | |
IGZGUYVVBABKOY-UHFFFAOYSA-N | |
1-iodo-2-(trifluoromethyl)benzene | |
67957 | |
99% |
Chemical Identifiers
444-29-1 | |
272.009 | |
IGZGUYVVBABKOY-UHFFFAOYSA-N | |
67957 | |
C1=CC=C(C(=C1)C(F)(F)F)I |
C7H4F3I | |
MFCD00001041 | |
2-iodobenzotrifluoride, 1-iodo-2-trifluoromethyl benzene, o-iodobenzotrifluoride, benzene, 1-iodo-2-trifluoromethyl, 2-iodobenztrifluoride, o-trifluoromethyliodobenzene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, iodobenzotrifluoride, 2-iodo-a,a,a-trifluorotoluene, 2-iodobenzotrifloride | |
1-iodo-2-(trifluoromethyl)benzene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA