missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodobenzonitrile 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC391800050

 View more versions of this product

Catalog No. AC391800050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
229.02
Yellow
100.0
2-Iodobenzonitrile
MFCD00079761
Glass bottle
C7H4IN
Solid
5g
50° to 54°C
4387-36-4
98.5
Authentic
IC6H4CN
99%

Chemical Identifiers

4387-36-4
100.0
C7H4IN
229.02
2-Iodobenzonitrile
98.5
99%
IC6H4CN
MFCD00079761
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Contact with acids liberates very toxic gas.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning