Learn More
2-Iodobenzoic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1056322
Description
Reagent for detection of sulfhydryl groups in proteins2-Iodobenzoic acid acts as a precursor in the preparation of 2-Iodoxybenzoic acid (IBX) and Dess-Martin periodinane, which finds application as an oxidizing agent in synthetic chemistry. It is also used in Suzuki reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReagent for detection of sulfhydryl groups in proteins2-Iodobenzoic acid acts as a precursor in the preparation of 2-Iodoxybenzoic acid (IBX) and Dess-Martin periodinane, which finds application as an oxidizing agent in synthetic chemistry. It is also used in Suzuki reaction.
Solubility
Soluble in dimethyl sulfoxide and methanol
Notes
Incompatible with oxidizing agents.
Specifications
2-Iodobenzoic acid | |
160°C to 164°C | |
C7H5IO2 | |
100 g | |
Light sensitive | |
o-iodobenzoic acid, benzoic acid, 2-iodo, iodobenzoic acid, benzoic acid, o-iodo, usaf ek-572, 2-iodo benzoic acid, 2-iodo-benzoic acid, benzoic acid, iodo, kyselina o-jodbenzoova, o-jod-benzoesaeure | |
CJNZAXGUTKBIHP-UHFFFAOYSA-N | |
2-iodobenzoic acid | |
6941 | |
248.02 |
88-67-5 | |
2.25 | |
MFCD00002419 | |
1861406 | |
14,5030 | |
Soluble in dimethyl sulfoxide and methanol | |
C1=CC=C(C(=C1)C(=O)O)I | |
248.019 | |
CHEBI:287979 | |
≥98% |
Chemical Identifiers
88-67-5 | |
248.019 | |
CJNZAXGUTKBIHP-UHFFFAOYSA-N | |
6941 | |
2-iodobenzoic acid |
C7H5IO2 | |
MFCD00002419 | |
o-iodobenzoic acid, benzoic acid, 2-iodo, iodobenzoic acid, benzoic acid, o-iodo, usaf ek-572, 2-iodo benzoic acid, 2-iodo-benzoic acid, benzoic acid, iodo, kyselina o-jodbenzoova, o-jod-benzoesaeure | |
CHEBI:287979 | |
C1=CC=C(C(=C1)C(=O)O)I |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
EINECSNumber : 201-850-7
RTECSNumber : DH2975000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only