missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Iodoanisole, 99%

Manufacturer:  Alfa Aesar™ A1015014

 View more versions of this product

Catalog No. AAA1015014



Description

Description

Applications
2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.

Solubility
Miscible with alcohol, chloroform and diethyl ether. Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications

Specifications

2-Iodoanisole
240°C to 242°C
529-28-2
MFCD00001039
14,5028
DVQWNQBEUKXONL-UHFFFAOYSA-N
1-iodo-2-methoxybenzene
68257
234.04
>110°C (230°F)
25g
C7H7IO
1860243
2-iodoanisole, o-iodoanisole, benzene, 1-iodo-2-methoxy, iodoanisole, 2-methoxyiodobenzene, o-anisyl iodide, 2-iodophenol methyl ether, anisole, o-iodo, 2-iodophenyl methyl ether, 1-iodo-2-methoxy-benzene
COC1=CC=CC=C1I
234.036
CHEBI:16355
99%

Chemical Identifiers

529-28-2
234.036
DVQWNQBEUKXONL-UHFFFAOYSA-N
68257
1-iodo-2-methoxybenzene
C7H7IO
MFCD00001039
2-iodoanisole, o-iodoanisole, benzene, 1-iodo-2-methoxy, iodoanisole, 2-methoxyiodobenzene, o-anisyl iodide, 2-iodophenol methyl ether, anisole, o-iodo, 2-iodophenyl methyl ether, 1-iodo-2-methoxy-benzene
CHEBI:16355
COC1=CC=CC=C1I
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents