Learn More
2-Iodoanisole, 97%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific 249680050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Iodoanisole | |
529-28-2 | |
100.0 | |
Yellow | |
>110°C | |
Authentic | |
IC6H4OCH3 | |
Glass bottle | |
1.6210 to 1.623 | |
1-iodo-2-methoxy-benzene, 2-iodoanisole, 2-iodophenol methyl ether, 2-iodophenyl methyl ether, 2-methoxyiodobenzene, anisole, o-iodo, benzene, 1-iodo-2-methoxy, iodoanisole, o-anisyl iodide, o-iodoanisole | |
DVQWNQBEUKXONL-UHFFFAOYSA-N | |
COC1=CC=CC=C1I | |
234.04 | |
CHEBI:16355 | |
Liquid |
97% | |
96.0 | |
1.7900g/mL | |
238°C to 240°C | |
96% min. (GC) | |
C7H7IO | |
MFCD00001039 | |
15, 5071 | |
5g | |
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether | |
1.79 | |
1-iodo-2-methoxybenzene | |
68257 | |
234.04 | |
97% |
Chemical Identifiers
529-28-2 | |
234.04 | |
DVQWNQBEUKXONL-UHFFFAOYSA-N | |
68257 | |
1-iodo-2-methoxybenzene |
C7H7IO | |
MFCD00001039 | |
1-iodo-2-methoxy-benzene, 2-iodoanisole, 2-iodophenol methyl ether, 2-iodophenyl methyl ether, 2-methoxyiodobenzene, anisole, o-iodo, benzene, 1-iodo-2-methoxy, iodoanisole, o-anisyl iodide, o-iodoanisole | |
CHEBI:16355 | |
COC1=CC=CC=C1I |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 208-456-4
TSCA : TSCA
Recommended Storage : Product may darken on storage
RUO – Research Use Only