missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodoanisole, 97%, ACROS Organics™

Chemical Identifiers

CAS 529-28-2
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.04
MDL Number MFCD00001039
InChI Key DVQWNQBEUKXONL-UHFFFAOYSA-N
Synonym 1-iodo-2-methoxy-benzene, 2-iodoanisole, 2-iodophenol methyl ether, 2-iodophenyl methyl ether, 2-methoxyiodobenzene, anisole, o-iodo, benzene, 1-iodo-2-methoxy, iodoanisole, o-anisyl iodide, o-iodoanisole
PubChem CID 68257
ChEBI CHEBI:16355
IUPAC Name 1-iodo-2-methoxybenzene
SMILES COC1=CC=CC=C1I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC249680050
SDS View Documents
Fisher Scientific Acros
249680050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC249680500
SDS View Documents
Fisher Scientific Acros
249680500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

529-28-2
234.04
DVQWNQBEUKXONL-UHFFFAOYSA-N
68257
1-iodo-2-methoxybenzene
C7H7IO
MFCD00001039
1-iodo-2-methoxy-benzene, 2-iodoanisole, 2-iodophenol methyl ether, 2-iodophenyl methyl ether, 2-methoxyiodobenzene, anisole, o-iodo, benzene, 1-iodo-2-methoxy, iodoanisole, o-anisyl iodide, o-iodoanisole
CHEBI:16355
COC1=CC=CC=C1I

Specifications

1.7900g/mL
Authentic
1.6210 to 1.623
238°C to 240°C
5g
96.0
96% min. (GC)
IC6H4OCH3
15, 5071
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
COC1=CC=CC=C1I
234.04
CHEBI:16355
Liquid
2-Iodoanisole, 97%
>110°C
Glass bottle
1.79
Yellow
529-28-2
100.0
C7H7IO
MFCD00001039
1-iodo-2-methoxy-benzene, 2-iodoanisole, 2-iodophenol methyl ether, 2-iodophenyl methyl ether, 2-methoxyiodobenzene, anisole, o-iodo, benzene, 1-iodo-2-methoxy, iodoanisole, o-anisyl iodide, o-iodoanisole
DVQWNQBEUKXONL-UHFFFAOYSA-N
1-iodo-2-methoxybenzene
68257
234.04
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 208-456-4

TSCA : TSCA

Recommended Storage : Product may darken on storage

SDS