missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodoanisole, 97%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 249680500

 View more versions of this product

Catalog No. AC249680500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Iodoanisole
>110°C
Glass bottle
1.79
Yellow
97%
96.0
96% min. (GC)
IC6H4OCH3
15, 5071
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
COC1=CC=CC=C1I
234.04
CHEBI:16355
Liquid
1.7900g/mL
Authentic
1.6210 to 1.623
238°C to 240°C
50g
529-28-2
100.0
C7H7IO
MFCD00001039
1-iodo-2-methoxy-benzene, 2-iodoanisole, 2-iodophenol methyl ether, 2-iodophenyl methyl ether, 2-methoxyiodobenzene, anisole, o-iodo, benzene, 1-iodo-2-methoxy, iodoanisole, o-anisyl iodide, o-iodoanisole
DVQWNQBEUKXONL-UHFFFAOYSA-N
1-iodo-2-methoxybenzene
68257
234.04
97%

Chemical Identifiers

529-28-2
234.04
DVQWNQBEUKXONL-UHFFFAOYSA-N
68257
1-iodo-2-methoxybenzene
C7H7IO
MFCD00001039
1-iodo-2-methoxy-benzene, 2-iodoanisole, 2-iodophenol methyl ether, 2-iodophenyl methyl ether, 2-methoxyiodobenzene, anisole, o-iodo, benzene, 1-iodo-2-methoxy, iodoanisole, o-anisyl iodide, o-iodoanisole
CHEBI:16355
COC1=CC=CC=C1I
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 208-456-4

TSCA : TSCA

Recommended Storage : Product may darken on storage

SDS Documents