missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodoaniline, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 122300050

 View more versions of this product

Catalog No. AC122300050



Description

Description

Specifications

Specifications

2-Iodoaniline
Authentic
Brown to Yellow
5g
97.5
98%
MFCD00007680
2-iodo aniline, 2-iodo-phenylamine, 2-iodobenzenamine, 2-iodophenylamine, aniline, 2-iodo, aniline, o-iodo, benzenamine, 2-iodo, benzenamine, iodo, o-aminoiodobenzene, o-iodoaniline
UBPDKIDWEADHPP-UHFFFAOYSA-N
2-iodoaniline
11995
Crystalline Needles
>110°C
Glass bottle
53°C to 58°C
615-43-0
100.0
C6H6IN
12, 669
Solubility in water: slightly soluble
NC1=CC=CC=C1I
219.03
219.02
98%

Chemical Identifiers

615-43-0
219.03
UBPDKIDWEADHPP-UHFFFAOYSA-N
11995
NC1=CC=CC=C1I
C6H6IN
MFCD00007680
2-iodo aniline, 2-iodo-phenylamine, 2-iodobenzenamine, 2-iodophenylamine, aniline, 2-iodo, aniline, o-iodo, benzenamine, 2-iodo, benzenamine, iodo, o-aminoiodobenzene, o-iodoaniline
2-iodoaniline
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 210-426-

SDS Documents
Product Certifications