missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodoadenosine, TCI America™

$145.14 - $1,397.94

Chemical Identifiers

CAS 35109-88-7
Molecular Formula C10H12IN5O4
Molecular Weight (g/mol) 393.141
MDL Number MFCD01320413
InChI Key MGEBVSZZNFOIRB-UUOKFMHZSA-N
Synonym 6-Amino-2-iodo-9-(beta-D-ribofuranosyl)purine
PubChem CID 169662
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-2-iodopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2N)I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I07595G View Documents TCI America
I07595G
5g Each for $1,397.94
 
I07591G View Documents TCI America
I07591G
1g Each for $432.38
 
I0759200MG View Documents TCI America
I0759200MG
200mg Each for $145.14
 
Specifications

Chemical Identifiers

35109-88-7
393.141
MGEBVSZZNFOIRB-UUOKFMHZSA-N
169662
C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2N)I
C10H12IN5O4
MFCD01320413
6-Amino-2-iodo-9-(beta-D-ribofuranosyl)purine
(2R,3R,4S,5R)-2-(6-amino-2-iodopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol

Specifications

White-Yellow
5g
C10H12IN5O4
6-Amino-2-iodo-9-(beta-D-ribofuranosyl)purine
C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2N)I
393.141
393.14
≥98.0% (HPLC,N)
144°C
35109-88-7
MFCD01320413
MGEBVSZZNFOIRB-UUOKFMHZSA-N
(2R,3R,4S,5R)-2-(6-amino-2-iodopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
169662
Crystal-Powder at 20°C
2-Iodoadenosine
Safety and Handling

Safety and Handling

TSCA : No