missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodo-5-nitrotoluene 96.0+%, TCI America™

$145.20 - $509.83

Chemical Identifiers

CAS 5326-38-5
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.034
MDL Number MFCD00051517
InChI Key ARJHCXYRCLMLQN-UHFFFAOYSA-N
Synonym 1-iodo-2-methyl-4-nitro-benzene, 2-iodo-5-nitrotoluene, 2-jod-5-nitro-1-methylbenzol, acmc-1amrp, benzene, 1-iodo-2-methyl-4-nitro, benzene,1-iodo-2-methyl-4-nitro
PubChem CID 79225
IUPAC Name 1-iodo-2-methyl-4-nitrobenzene
SMILES CC1=C(C=CC(=C1)[N+](=O)[O-])I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09925G View Documents Tci America
I09925G
5g Each for $145.20
 
I099225G View Documents Tci America
I099225G
25g Each for $509.83
 
Specifications

Chemical Identifiers

5326-38-5
263.034
ARJHCXYRCLMLQN-UHFFFAOYSA-N
79225
CC1=C(C=CC(=C1)[N+](=O)[O-])I
C7H6INO2
MFCD00051517
1-iodo-2-methyl-4-nitro-benzene, 2-iodo-5-nitrotoluene, 2-jod-5-nitro-1-methylbenzol, acmc-1amrp, benzene, 1-iodo-2-methyl-4-nitro, benzene,1-iodo-2-methyl-4-nitro
1-iodo-2-methyl-4-nitrobenzene

Specifications

Undesignated
5g
C7H6INO2
1-iodo-2-methyl-4-nitro-benzene, 2-iodo-5-nitrotoluene, 2-jod-5-nitro-1-methylbenzol, acmc-1amrp, benzene, 1-iodo-2-methyl-4-nitro, benzene,1-iodo-2-methyl-4-nitro
CC1=C(C=CC(=C1)[N+](=O)[O-])I
263.034
263.03
≥96.0% (GC)
102°C
5326-38-5
MFCD00051517
ARJHCXYRCLMLQN-UHFFFAOYSA-N
1-iodo-2-methyl-4-nitrobenzene
79225
Crystal-Powder at 20°C
2-Iodo-5-nitrotoluene
Safety and Handling

Safety and Handling

TSCA : No