Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Iodo-5-nitrotoluene, 98%

$289.30 - $289.30

Chemical Identifiers

Chemical Name or Material 2-Iodo-5-nitrotoluene
CAS 5326-38-5
Molecular Formula C7H6INO2
Formula Weight 263.03
MDL Number MFCD00051517
Beilstein 2438587
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0784214

  SDS
alfa aesar™
L0784214
25g Each for $289.30
Description & Specifications

Specifications

Formula Weight 263.03
CAS 5326-38-5
Chemical Name or Material 2-Iodo-5-nitrotoluene
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00051517
Molecular Formula C7H6INO2
Assay 98%
Health Hazard 1 Warning
Beilstein 2438587
Quantity 25g
Melting Point 99° to 104°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.