missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodo-5-methylthiophene, 97%, ACROS Organics™

Chemical Identifiers

CAS 16494-36-3
Molecular Formula C5H5IS
Molecular Weight (g/mol) 224.06
InChI Key NAZNQEXKAPLVKC-UHFFFAOYSA-N
Synonym 2-iodo-5-methyl thiophene, 2-iodo-5-methyl-thiophene, 2-iodo-5-methylthiophene, acmc-1bs6v, bidd:gt0402, pubchem7417, thiophene, 2-iodo-5-methyl
PubChem CID 140096
IUPAC Name 2-iodo-5-methylthiophene
SMILES CC1=CC=C(S1)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC439270010
SDS View Documents
Thermo Fisher Scientific
AC439270010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC439270050
SDS View Documents
Thermo Fisher Scientific
AC439270050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

16494-36-3
224.06
2-iodo-5-methyl thiophene, 2-iodo-5-methyl-thiophene, 2-iodo-5-methylthiophene, acmc-1bs6v, bidd:gt0402, pubchem7417, thiophene, 2-iodo-5-methyl
2-iodo-5-methylthiophene
C5H5IS
NAZNQEXKAPLVKC-UHFFFAOYSA-N
140096
CC1=CC=C(S1)I

Specifications

1.8520g/mL
Authentic
1.852
Orange-Red to Yellow
1g
96.0
96.5% min. (GC)
2-iodo-5-methyl thiophene, 2-iodo-5-methyl-thiophene, 2-iodo-5-methylthiophene, acmc-1bs6v, bidd:gt0402, pubchem7417, thiophene, 2-iodo-5-methyl
CC1=CC=C(S1)I
224.06
224.06
97%
89°C
Glass Bottle
81°C to 83°C (10.0mmHg)
-25°C to 28°C
16494-36-3
100.0
C5H5IS
NAZNQEXKAPLVKC-UHFFFAOYSA-N
2-iodo-5-methylthiophene
140096
Liquid
2-Iodo-5-methylthiophene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS