missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodo-5-methylthiophene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439270050

 View more versions of this product

Catalog No. AC439270050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Iodo-5-methylthiophene
89°C
Glass bottle
81°C to 83°C (10.0mmHg)
-25°C to 28°C
16494-36-3
100.0
C5H5IS
NAZNQEXKAPLVKC-UHFFFAOYSA-N
2-iodo-5-methylthiophene
140096
Liquid
1.8520g/mL
Authentic
1.852
Orange-Red to Yellow
5g
96.0
96.5% min. (GC)
2-iodo-5-methyl thiophene, 2-iodo-5-methyl-thiophene, 2-iodo-5-methylthiophene, acmc-1bs6v, bidd:gt0402, pubchem7417, thiophene, 2-iodo-5-methyl
CC1=CC=C(S1)I
224.06
224.06
97%

Chemical Identifiers

16494-36-3
224.06
2-iodo-5-methyl thiophene, 2-iodo-5-methyl-thiophene, 2-iodo-5-methylthiophene, acmc-1bs6v, bidd:gt0402, pubchem7417, thiophene, 2-iodo-5-methyl
2-iodo-5-methylthiophene
C5H5IS
NAZNQEXKAPLVKC-UHFFFAOYSA-N
140096
CC1=CC=C(S1)I
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents