Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Iodo-4-(trifluoromethyl)benzoic acid, 98%

$931.27 - $931.27

Chemical Identifiers

Chemical Name or Material 2-Iodo-4-(trifluoromethyl)benzoic acid
CAS 54507-44-7
Molecular Formula C8H4F3IO2
Formula Weight 316.02
MDL Number MFCD06408817
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3283906

 
alfa aesar™
H3283906
5g Each for $931.27
Description & Specifications

Specifications

Formula Weight 316.02
Chemical Name or Material 2-Iodo-4-(trifluoromethyl)benzoic acid
Assay 98%
Molecular Formula C8H4F3IO2
MDL Number MFCD06408817
CAS 54507-44-7
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
The 10-chloro-8-fluoro-10, 11-dihydro-2-trifluoromethyldibenzothiepin used as the starting material can be prepared starting from 2-iodo--4-trifluoromethyl-benzoic acid and 4-fluoro-thiophenol.

Solubility
Slightly soluble.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.