missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodo-3-nitrotoluene, TCI America™

$125.28 - $432.38

Chemical Identifiers

CAS 6277-17-4
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.034
MDL Number MFCD00091040
InChI Key AKYPSJARSQSCHJ-UHFFFAOYSA-N
Synonym 2-Iodo-1-methyl-3-nitrobenzene
PubChem CID 234781
IUPAC Name 2-iodo-1-methyl-3-nitrobenzene
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I07995G View Documents TCI America
I07995G
5g Each for $125.28
 
I079925G View Documents TCI America
I079925G
25g Each for $432.38
 
Specifications

Chemical Identifiers

6277-17-4
263.034
AKYPSJARSQSCHJ-UHFFFAOYSA-N
234781
CC1=C(C(=CC=C1)[N+](=O)[O-])I
C7H6INO2
MFCD00091040
2-Iodo-1-methyl-3-nitrobenzene
2-iodo-1-methyl-3-nitrobenzene

Specifications

Yellow
5g
C7H6INO2
2-Iodo-1-methyl-3-nitrobenzene
CC1=C(C(=CC=C1)[N+](=O)[O-])I
263.034
263.03
≥98.0% (GC)
67°C
6277-17-4
MFCD00091040
AKYPSJARSQSCHJ-UHFFFAOYSA-N
2-iodo-1-methyl-3-nitrobenzene
234781
Crystal-Powder at 20°C
2-Iodo-3-nitrotoluene
Safety and Handling

Safety and Handling

TSCA : No