Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Iodo-3-methylpyridine, 97%

$81.58 - $295.54

Chemical Identifiers

Chemical Name or Material 2-Iodo-3-methylpyridine
CAS 22282-58-2
Molecular Formula C6H6IN
Formula Weight 219.03
MDL Number MFCD04039350
Beilstein 5328169
Synonym 2-Iodo-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2004703

 
alfa aesar™
L2004703
1g Each for $81.58

AAL2004706

 
alfa aesar™
L2004706
5g Each for $295.54
Description & Specifications

Specifications

Formula Weight 219.03
Assay 97%
Beilstein 5328169
Boiling Point 80°C (0.3mmHg)
Chemical Name or Material 2-Iodo-3-methylpyridine
Molecular Formula C6H6IN
MDL Number MFCD04039350
CAS 22282-58-2
Synonym 2-Iodo-3-picoline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C