Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Iodo-3-methoxypyridine, 97%

$59.89 - $846.61

Chemical Identifiers

Chemical Name or Material 2-Iodo-3-methoxypyridine
CAS 93560-55-5
Molecular Formula C6H6INO
Formula Weight 235.03
MDL Number MFCD00234181
Beilstein 115569
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1986803

  SDS
alfa aesar™
L1986803
1g Each for $59.89

AAL1986806

  SDS
alfa aesar™
L1986806
5g Each for $210.88

AAL1986814

  SDS
alfa aesar™
L1986814
25g Each for $846.61
Description & Specifications

Specifications

Formula Weight 235.03
Chemical Name or Material 2-Iodo-3-methoxypyridine
Assay 97%
Molecular Formula C6H6INO
MDL Number MFCD00234181
CAS 93560-55-5
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 115569
Melting Point 57° to 61°C
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.