Please login to your online account to display your discounted pricing

2-Iodo-3-hydroxypyridine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Hydroxy-2-iodopyridine
CAS 40263-57-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H4INO
Formula Weight 220.99
Synonym 2-Iodo-3-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC436390050

  SDS
thermo fisher scientific
AC436390050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC436390010

  SDS
thermo fisher scientific
AC436390010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 220.99
Physical Form Crystalline Powder
Assay ≥96.0%
Quantity 5g
Color White to tan
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 1 Warning
Assay Percent Range 97%
CAS 40263-57-8
Chemical Name or Material 3-Hydroxy-2-iodopyridine
Molecular Formula C5H4INO
Elemental Analysis 26.08 to 28.25% C, 6.09 to 6.59% N
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 2-Iodo-3-pyridinol
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.