missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Hydroxy-2-iodopyridine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436390050
Specifications
3-Hydroxy-2-iodopyridine | |
Authentic | |
Tan to White | |
5g | |
96.0 | |
96% min. (HPLC) | |
2-iodo-3-hydroxy-pyridine, 2-iodo-3-hydroxypyridin, 2-iodo-3-hydroxypyridine, 2-iodo-3-pyridinol, 2-iodo-pyridin-3-ol, 2-iodopyridine-3-ol, 2-iodpyridin-3-ol, 3-hydroxy-2-iodopyridine, 3-pyridinol, 2-iodo, acmc-209jcf | |
C1=CC(=C(N=C1)I)O | |
220.99 | |
220.99 | |
97% |
26.08 to 28.25% C, 6.09 to 6.59% N | |
Glass bottle | |
191.4°C to 193.2°C | |
40263-57-8 | |
100.0 | |
C5H4INO | |
HJBGMPCMSWJZNH-UHFFFAOYSA-N | |
2-iodopyridin-3-ol | |
97179 | |
Crystalline Powder |
Chemical Identifiers
40263-57-8 | |
220.99 | |
2-iodo-3-hydroxy-pyridine, 2-iodo-3-hydroxypyridin, 2-iodo-3-hydroxypyridine, 2-iodo-3-pyridinol, 2-iodo-pyridin-3-ol, 2-iodopyridine-3-ol, 2-iodpyridin-3-ol, 3-hydroxy-2-iodopyridine, 3-pyridinol, 2-iodo, acmc-209jcf | |
2-iodopyridin-3-ol |
C5H4INO | |
HJBGMPCMSWJZNH-UHFFFAOYSA-N | |
97179 | |
C1=CC(=C(N=C1)I)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 254-864-