Learn More
2-Iodo-2-methylpropane, 95%, stab. with copper powder, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0278622
Description
2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.
Solubility
Miscible with ethanol and ether.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
Specifications
2-Iodo-2-methylpropane | |
558-17-8 | |
1.547 | |
7°C (44°F) | |
C4H9I | |
MFCD00001068 | |
UN2391 | |
Light sensitive | |
Miscible with ethanol and ether. | |
CC(C)(C)I | |
184.02 | |
184.02 |
stabilized with copper powder | |
-38°C | |
99°C to 100°C | |
Recognizable | |
1.491 | |
100 g | |
505950 | |
tert-butyl iodide, propane, 2-iodo-2-methyl, trimethyliodomethane, t-butyl iodide, unii-e5hy19g10p, 2-jod-2-methylpropan, sjphavimup@, tert-c4h9i, 2-iodo-2-methyl propane, 2-iodo-2-methyl-propane | |
ANGGPYSFTXVERY-UHFFFAOYSA-N | |
2-iodo-2-methylpropane | |
11206 | |
95% |
Chemical Identifiers
558-17-8 | |
184.02 | |
ANGGPYSFTXVERY-UHFFFAOYSA-N | |
11206 | |
CC(C)(C)I |
C4H9I | |
MFCD00001068 | |
tert-butyl iodide, propane, 2-iodo-2-methyl, trimethyliodomethane, t-butyl iodide, unii-e5hy19g10p, 2-jod-2-methylpropan, sjphavimup@, tert-c4h9i, 2-iodo-2-methyl propane, 2-iodo-2-methyl-propane | |
2-iodo-2-methylpropane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: IODOMETHYLPROPANES
EINECSNumber : 209-190-1
RTECSNumber : TZ4251000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only