Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Iodo-1-tritylimidazole, 97%

$78.50 - $232.43

Chemical Identifiers

Chemical Name or Material 2-Iodo-1-tritylimidazole
CAS 67478-46-0
Molecular Formula C22H17IN2
Formula Weight 436.29
MDL Number MFCD02179529
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5187103

  SDS
alfa aesar™
H5187103
1g Each for $232.43

AAH51871MD

  SDS
alfa aesar™
H51871MD
250mg Each for $78.50
Description & Specifications

Specifications

Formula Weight 436.29
Chemical Name or Material 2-Iodo-1-tritylimidazole
Assay 97%
Molecular Formula C22H17IN2
MDL Number MFCD02179529
CAS 67478-46-0
Melting Point 170° to 171°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.