Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Indanylboronic acid diethanolamine ester, 98%

$75.78 - $75.78

Chemical Identifiers

Chemical Name or Material 2-Indanylboronic acid diethanolamine ester
CAS 501014-44-4
Molecular Formula C13H18BNO2
Formula Weight 231.1
MDL Number MFCD03788732
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19574MC

  SDS
alfa aesar™
L19574MC
100mg Each for $75.78
Description & Specifications

Specifications

Formula Weight 231.1
CAS 501014-44-4
MDL Number MFCD03788732
Health Hazard 1 Warning
Melting Point 213° to 219°C
Quantity 100mg
Assay 98%
Molecular Formula C13H18BNO2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Indanylboronic acid diethanolamine ester
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.