Learn More
2-Indanylacetic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1070003
Description
2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
2-Indanylacetic acid | |
91°C to 93°C | |
MFCD00082653 | |
2616409 | |
Insoluble in water. | |
OC(=O)CC1CC2=CC=CC=C2C1 | |
176.22 | |
176.22 |
37868-26-1 | |
C11H12O2 | |
1 g | |
2-2,3-dihydro-1h-inden-2-yl acetic acid, indan-2-yl-acetic acid, 1h-indene-2-acetic acid, 2,3-dihydro, 2,3-dihydro-1h-inden-2-ylacetic acid, 2,3-dihydro-1h-indene-2-acetic acid, 2-indaneacetic acid, indan-2-acetic acid, pubchem16311, indan-2-yl acetic acid | |
TULDPXYHBFBRGW-UHFFFAOYSA-N | |
2-(2,3-dihydro-1H-inden-2-yl)acetic acid | |
5174955 | |
99% |
Chemical Identifiers
37868-26-1 | |
176.22 | |
TULDPXYHBFBRGW-UHFFFAOYSA-N | |
5174955 | |
OC(=O)CC1CC2=CC=CC=C2C1 |
C11H12O2 | |
MFCD00082653 | |
2-2,3-dihydro-1h-inden-2-yl acetic acid, indan-2-yl-acetic acid, 1h-indene-2-acetic acid, 2,3-dihydro, 2,3-dihydro-1h-inden-2-ylacetic acid, 2,3-dihydro-1h-indene-2-acetic acid, 2-indaneacetic acid, indan-2-acetic acid, pubchem16311, indan-2-yl acetic acid | |
2-(2,3-dihydro-1H-inden-2-yl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only