Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Indanyl p-toluenesulfonate, 98+%

$48.34 - $172.41

Chemical Identifiers

Chemical Name or Material 2-Indanyl p-toluenesulfonate
CAS 17783-69-6
Molecular Formula C16H16O3S
Formula Weight 288.37
MDL Number MFCD00082747
Beilstein 2147482
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1291606

 
alfa aesar™
L1291606
5g Each for $48.34

AAL1291614

 
alfa aesar™
L1291614
25g Each for $172.41
Description & Specifications

Specifications

Formula Weight 288.37
Chemical Name or Material 2-Indanyl p-toluenesulfonate
Assay ≥98%
Molecular Formula C16H16O3S
MDL Number MFCD00082747
CAS 17783-69-6
Beilstein 2147482
Melting Point 118° to 121°C
Quantity 5g
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C