missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Indanyl p-toluenesulfonate, 98+%

Manufacturer:  Alfa Aesar™ L1291614

 View more versions of this product

Catalog No. AAL1291614



Specifications

Specifications

2-Indanyl p-toluenesulfonate
25g
C16H16O3S
2147482
QUYCPYFNAZEWOT-UHFFFAOYSA-N
2,3-dihydro-1H-inden-2-yl 4-methylbenzenesulfonate
7021474
≥98%
118°C to 121°C
17783-69-6
MFCD00082747
2-indanyl p-toluenesulfonate, 2-indanyl p-toluenesulphonate, indan-2-yl tosylate, 2-indanyl para-toluenesulfonate, indan-2-yl 4-methylbenzenesulfonate, p-toluenesulfonic acid indan-2-yl ester
CC1=CC=C(C=C1)S(=O)(=O)OC2CC3=CC=CC=C3C2
288.361
288.37

Chemical Identifiers

17783-69-6
288.361
QUYCPYFNAZEWOT-UHFFFAOYSA-N
7021474
CC1=CC=C(C=C1)S(=O)(=O)OC2CC3=CC=CC=C3C2
C16H16O3S
MFCD00082747
2-indanyl p-toluenesulfonate, 2-indanyl p-toluenesulphonate, indan-2-yl tosylate, 2-indanyl para-toluenesulfonate, indan-2-yl 4-methylbenzenesulfonate, p-toluenesulfonic acid indan-2-yl ester
2,3-dihydro-1H-inden-2-yl 4-methylbenzenesulfonate
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

Documents