Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Indanone, 98%

$257.66 - $914.61

Chemical Identifiers

Chemical Name or Material 2-Indanone
CAS 615-13-4
Molecular Formula C9H8O
Formula Weight 132.16
MDL Number MFCD00003792
Beilstein 636550
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1257414

  SDS
alfa aesar™
A1257414
25g Each for $257.66

AAA1257422

  SDS
alfa aesar™
A1257422
100g Each for $914.61

AAA1257406

  SDS
alfa aesar™
A1257406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Indanone
CAS 615-13-4
Recommended Storage 2° to 8°C
RTECS Number NK7535500
TSCA TSCA
Assay 98%
Molecular Formula C9H8O
Formula Weight 132.16
MDL Number MFCD00003792
Beilstein 636550
Boiling Point 218°C (dec.)
Melting Point 52° to 56°C
Flash Point 100°C (212°F)
Quantity 25g

Applications
2-Indanone undergoes TiCl4-Mg mediated coupling with CHBr3 to yield dibromomethyl carbinol. It reacts with 5,5-dimethyl-3-pyrazolidinone to yield 5,5-dimethyl-2-(1H-indenyl-2)-3-pyrazolidinone. On photolysis by 266-nm one-photon excitation yields o-xylylene. It was used as starting reagent in the synthesis of indene-fused porphyrins.

Solubility
Insoluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents. Incompatible with oxidizing agents and bases.

Safety and Handling

2° to 8°C