missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Indanone, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 164110500

 View more versions of this product

Catalog No. AC164110500



Specifications

Specifications

2-Indanone
Authentic
Brown-Yellow to Yellow
50g
615-13-4
100.0
C9H8O
07, 363
Solubility in water: insoluble
C1C(=O)CC2=CC=CC=C21
132.16
CHEBI:27930
Crystals or Powder
100°C
Glass bottle
50.0°C to 57.0°C
97%
96.0
96% min. (GC)
MFCD00003792
1,3-dihydro-2h-inden-2-one
UMJJFEIKYGFCAT-UHFFFAOYSA-N
1,3-dihydroinden-2-one
11983
132.16
97%

Chemical Identifiers

615-13-4
132.16
UMJJFEIKYGFCAT-UHFFFAOYSA-N
11983
1,3-dihydroinden-2-one
C9H8O
MFCD00003792
1,3-dihydro-2h-inden-2-one
CHEBI:27930
C1C(=O)CC2=CC=CC=C21
Safety and Handling

Safety and Handling

EINECSNumber : 210-410-3

RTECSNumber : NK7535500

TSCA : TSCA

SDS Documents