missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Indanol, 99%

$75.16 - $203.76

Chemical Identifiers

CAS 4254-29-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00003800
InChI Key KMGCKSAIIHOKCX-UHFFFAOYSA-N
Synonym 1h-inden-2-ol, 2,3-dihydro, 2-hydroxyhydrindene, 2-hydroxyindan, 2-hydroxyindane, 2-indanol, 2-indanol;, acmc-1asoy, indan-2-ol, pubchem9652
PubChem CID 77936
IUPAC Name 2,3-dihydro-1H-inden-2-ol
SMILES C1C(CC2=CC=CC=C21)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0176614
View Documents
Alfa Aesar™
L0176614
25g
Each for $75.16
 
AAL0176622
View Documents
Alfa Aesar™
L0176622
100g
Each for $203.76
 
Description

Description

Specifications

Chemical Identifiers

4254-29-9
134.178
KMGCKSAIIHOKCX-UHFFFAOYSA-N
77936
C1C(CC2=CC=CC=C21)O
C9H10O
MFCD00003800
1h-inden-2-ol, 2,3-dihydro, 2-hydroxyhydrindene, 2-hydroxyindan, 2-hydroxyindane, 2-indanol, 2-indanol;, acmc-1asoy, indan-2-ol, pubchem9652
2,3-dihydro-1H-inden-2-ol

Specifications

109°C (228°F)
67°C to 72°C
7
C9H10O
1862567
KMGCKSAIIHOKCX-UHFFFAOYSA-N
2,3-dihydro-1H-inden-2-ol
77936
99%
120°C to 123°C (12mmHg)
25g
4254-29-9
MFCD00003800
1h-inden-2-ol, 2,3-dihydro, 2-hydroxyhydrindene, 2-hydroxyindan, 2-hydroxyindane, 2-indanol, 2-indanol;, acmc-1asoy, indan-2-ol, pubchem9652
C1C(CC2=CC=CC=C21)O
134.178
134.18
2-Indanol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 224-230-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS