Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Indanol, 99%


Quantity 100g

Manufacturer: alfa aesar™  L0176622

 View more versions of this product

Catalog No. AAL0176622

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 134.18
Molecular Formula C9H10O
MDL Number MFCD00003800
Assay 99%
CAS 4254-29-9
Synonym 2-Hydroxyindane
Health Hazard 1 Danger
Melting Point 67° to 72°C
pH 7
Quantity 100g
Health Hazard 2 GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Indanol
Beilstein 1862567
Boiling Point 120° to 123°C (12mmHg)
Flash Point 109°C (228°F)

Specifications

Chemical Name or Material 2-Indanol
CAS 4254-29-9
Molecular Formula C9H10O
Formula Weight 134.18
MDL Number MFCD00003800
Beilstein 1862567
Synonym 2-Hydroxyindane
Accessories

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

SDS & Safety Documents