Learn More
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 450182500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Iminothiolane Hydrochloride | |
97.5% min. (Argentometry) | |
C4H7NS·ClH | |
2-iminothiolane hydrochloride, 2-iminothiolane hcl, thiolan-2-imine hydrochloride, dihydrothiophen-2 3h-imine hydrochloride, 2 3h-thiophenimine, dihydro-, hydrochloride, traut's reagent, c4h7ns.clh, traut eo 1/4 a, 2-thiolanimine hydrochloride, 2 3h-thiophenimine, hydrochloride | |
C1CC(=N)SC1.Cl | |
137.63 | |
137.63 |
4781-83-3 | |
Glass Bottle | |
250 mg | |
ATGUDZODTABURZ-UHFFFAOYSA-N | |
thiolan-2-imine;hydrochloride | |
13166855 | |
98% |
Chemical Identifiers
4781-83-3 | |
137.63 | |
2-iminothiolane hydrochloride, 2-iminothiolane hcl, thiolan-2-imine hydrochloride, dihydrothiophen-2 3h-imine hydrochloride, 2 3h-thiophenimine, dihydro-, hydrochloride, traut's reagent, c4h7ns.clh, traut eo 1/4 a, 2-thiolanimine hydrochloride, 2 3h-thiophenimine, hydrochloride | |
thiolan-2-imine;hydrochloride |
C4H7NS·ClH | |
ATGUDZODTABURZ-UHFFFAOYSA-N | |
13166855 | |
C1CC(=N)SC1.Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only