Please login to your online account to display your discounted pricing

2-Iminopiperidine hydrochloride, Tocris


Selective iNOS inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS095110

 View more versions of this product

Catalog No. 095110

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 134.61
Chemical Name or Material 2-Iminopiperidine hydrochloride
Assay Percent Range >98%
Molecular Formula C5H10N2·HCl
CAS 16011-96-4
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material 2-Iminopiperidine hydrochloride
Assay Percent Range >98%
Molecular Formula C5H10N2·HCl
Formula Weight 134.61
Accessories

Store at Room Temperature