Please login to your online account to display your discounted pricing

2-Iminopiperidine hydrochloride, Tocris

Selective iNOS inhibitor

$133.98 - $567.83

Chemical Identifiers

Chemical Name or Material 2-Iminopiperidine hydrochloride
Assay Percent Range >98%
Molecular Formula C5H10N2·HCl
Formula Weight 134.61
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

095110

 
r & d systems
RDS095110
10mg Each for $133.98

095150

 
r & d systems
RDS095150
50mg Each for $567.83
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 134.61
Chemical Name or Material 2-Iminopiperidine hydrochloride
Assay Percent Range >98%
Molecular Formula C5H10N2·HCl
CAS 16011-96-4
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature