Learn More
2-Imidazolidone hemihydrate, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 122040250
Specifications
2-Imidazolidone hemihydrate | |
265°C | |
Plastic bottle | |
>100.0°C | |
58.0°C | |
121325-67-5,120-93-4 | |
100.0 | |
0·5 H2O | |
14, 4914 | |
Solubility in water: 950g/L (20°C). Other solubilities: 1800g/L water (40°C) | |
C1CNC(=O)N1.C1CNC(=O)N1.O | |
95.1 | |
95.1 | |
99+% |
265g/mL | |
Authentic | |
13% max. | |
White to Yellow | |
25g | |
99.0 | |
99% min. (GC) | |
MFCD02662355 | |
2-imidazolidinone hemihydrate, 2-imidazolidinone, hydrate 2:1, 2-imidazolidone hemihydrate, bis imidazolidin-2-one hydrate, bis imidazolidinone hydrate, imidazolidin-2-one hydrate, imidazolidin-2-one, imidazolidin-2-one, hydrate | |
KPRJGGOOWATRNT-UHFFFAOYSA-N | |
imidazolidin-2-one;hydrate | |
2723651 | |
Adhering Crystals or Crystalline Powder |
Chemical Identifiers
121325-67-5 | |
95.1 | |
KPRJGGOOWATRNT-UHFFFAOYSA-N | |
2723651 | |
C1CNC(=O)N1.C1CNC(=O)N1.O |
0·5 H2O | |
MFCD02662355 | |
2-imidazolidinone hemihydrate, 2-imidazolidinone, hydrate 2:1, 2-imidazolidone hemihydrate, bis imidazolidin-2-one hydrate, bis imidazolidinone hydrate, imidazolidin-2-one hydrate, imidazolidin-2-one, imidazolidin-2-one, hydrate | |
imidazolidin-2-one;hydrate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Suspected of damaging fertility.
GHS P Statement:
Use personal protective equipment as required.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning