missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Imidazolecarboxylic Acid Hydrate, TCI America™

$79.45 - $319.31

Chemical Identifiers

CAS 16042-25-4
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.088
MDL Number MFCD00270816
InChI Key KYWMCFOWDYFYLV-UHFFFAOYSA-N
Synonym 2-imidazolecarboxylic acid, imidazole-2-carboxylic acid, unii-o9g6515vc1, 1h-imidazolecarboxylic acid, 2-imidazolecarboxylicacid, 2-imidazocarboxylic acid, 1h-imidazole-2-carboxylicacid, imidazolecarboxylic, acmc-1avrt, 2-carboxy-1h-imidazole
PubChem CID 574321
IUPAC Name 1H-imidazole-2-carboxylic acid
SMILES C1=CN=C(N1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I08671G View Documents TCI America
I08671G
1g Each for $79.45
 
I08675G View Documents TCI America
I08675G
5g Each for $319.31
 
Specifications

Chemical Identifiers

16042-25-4
112.088
KYWMCFOWDYFYLV-UHFFFAOYSA-N
574321
C1=CN=C(N1)C(=O)O
C4H4N2O2
MFCD00270816
2-imidazolecarboxylic acid, imidazole-2-carboxylic acid, unii-o9g6515vc1, 1h-imidazolecarboxylic acid, 2-imidazolecarboxylicacid, 2-imidazocarboxylic acid, 1h-imidazole-2-carboxylicacid, imidazolecarboxylic, acmc-1avrt, 2-carboxy-1h-imidazole
1H-imidazole-2-carboxylic acid

Specifications

Blue-White
1g
C4H4N2O2
2-imidazolecarboxylic acid, imidazole-2-carboxylic acid, unii-o9g6515vc1, 1h-imidazolecarboxylic acid, 2-imidazolecarboxylicacid, 2-imidazocarboxylic acid, 1h-imidazole-2-carboxylicacid, imidazolecarboxylic, acmc-1avrt, 2-carboxy-1h-imidazole
C1=CN=C(N1)C(=O)O
112.088
112.09
≥98.0% (HPLC)
157°C
16042-25-4
MFCD00270816
KYWMCFOWDYFYLV-UHFFFAOYSA-N
1H-imidazole-2-carboxylic acid
574321
Crystal-Powder at 20°C
2-Imidazolecarboxylic Acid Hydrate
Safety and Handling

Safety and Handling

TSCA : No