Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxythiophenol, 97%

Chemical Identifiers

Chemical Name or Material 2-Hydroxythiophenol
CAS 1121-24-0
Molecular Formula C6H6OS
Formula Weight 126.18
MDL Number MFCD00040447
UN Number UN2922
Beilstein 1280884
Synonym 2-Mercaptophenol; Monothiocatechol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1251903

 
alfa aesar™
L1251903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1251906

 
alfa aesar™
L1251906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.18
Chemical Name or Material 2-Hydroxythiophenol
Molecular Formula C6H6OS
MDL Number MFCD00040447
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay 97%
UN Number UN2922
Beilstein 1280884
Synonym 2-Mercaptophenol; Monothiocatechol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
RTECS Number SL7375000
Flash Point 97°C (206°F)
Odor Unpleasant
Boiling Point 217°C
Melting Point 19°C
Quantity 1g
CAS 1121-24-0
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.