missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxythiophenol, 97%

Manufacturer:  Alfa Aesar™ L1251906

 View more versions of this product

Catalog No. AAL1251906



Specifications

Specifications

2-Hydroxythiophenol
Unpleasant
19°C
1121-24-0
MFCD00040447
1280884
VMKYTRPNOVFCGZ-UHFFFAOYSA-N
2-sulfanylphenol
70718
97%
97°C (206°F)
217°C
5g
C6H6OS
UN2922
2-hydroxythiophenol, 2-mercaptophenol, o-mercaptophenol, phenol, 2-mercapto, 2-hydroxy thiophenol, 2-hydroxybenzenethiol, o-hydroxythiophenol, o-hydroxybenzenothiol, 2-hydroxybenzenothiol, mercaptophenol
C1=CC=C(C(=C1)O)S
126.173
126.18

Chemical Identifiers

1121-24-0
126.173
VMKYTRPNOVFCGZ-UHFFFAOYSA-N
70718
C1=CC=C(C(=C1)O)S
C6H6OS
MFCD00040447
2-hydroxythiophenol, 2-mercaptophenol, o-mercaptophenol, phenol, 2-mercapto, 2-hydroxy thiophenol, 2-hydroxybenzenethiol, o-hydroxythiophenol, o-hydroxybenzenothiol, 2-hydroxybenzenothiol, mercaptophenol
2-sulfanylphenol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : SL7375000

Documents