Please login to your online account to display your discounted pricing

2-Hydroxysaclofen, Tocris

Selective GABAB antagonist, more potent than saclofen

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material 2-Hydroxysaclofen
Assay Percent Range >98%
Molecular Formula C9H12ClNO4S
Formula Weight 265.71
Synonym (RS)-3-Amino-2-(4-chlorophenyl)-2-hydroxypropyl-sulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

024550

 
r & d systems
RDS024550
50mg Each for $997.23

024510

 
r & d systems
RDS024510
10mg Each for $244.81
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 265.71
Chemical Name or Material 2-Hydroxysaclofen
Assay Percent Range >98%
Molecular Formula C9H12ClNO4S
Recommended Storage Store at Room Temperature
CAS 117354-64-0
Synonym (RS)-3-Amino-2-(4-chlorophenyl)-2-hydroxypropyl-sulfonic acid
Safety and Handling

Store at Room Temperature