missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxyquinoxaline, TCI America™

$51.95 - $357.50

Chemical Identifiers

CAS 1196-57-2
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD00006722
InChI Key FFRYUAVNPBUEIC-UHFFFAOYSA-N
Synonym 2-hydroxyquinoxaline, quinoxalin-2-ol, 2-quinoxalinol, 2 1h-quinoxalinone, quinoxalin-2 1h-one, quinoxalin-2-one, 2-quinoxalinone, 3-quinoxalinone, 2-quinoxalone, 1,2-dihydroquinoxalin-2-one
PubChem CID 14526
ChEBI CHEBI:38890
IUPAC Name 1H-quinoxalin-2-one
SMILES C1=CC=C2C(=C1)NC(=O)C=N2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H0591500G View Documents TCI America
H0591500G
500g Each for $357.50
 
H0591100G View Documents TCI America
H0591100G
100g Each for $126.81
 
H059125G View Documents TCI America
H059125G
25g Each for $51.95
 
Specifications

Chemical Identifiers

1196-57-2
146.149
FFRYUAVNPBUEIC-UHFFFAOYSA-N
14526
1H-quinoxalin-2-one
C8H6N2O
MFCD00006722
2-hydroxyquinoxaline, quinoxalin-2-ol, 2-quinoxalinol, 2 1h-quinoxalinone, quinoxalin-2 1h-one, quinoxalin-2-one, 2-quinoxalinone, 3-quinoxalinone, 2-quinoxalone, 1,2-dihydroquinoxalin-2-one
CHEBI:38890
C1=CC=C2C(=C1)NC(=O)C=N2

Specifications

Undesignated
500g
C8H6N2O
2-hydroxyquinoxaline, quinoxalin-2-ol, 2-quinoxalinol, 2 1h-quinoxalinone, quinoxalin-2 1h-one, quinoxalin-2-one, 2-quinoxalinone, 3-quinoxalinone, 2-quinoxalone, 1,2-dihydroquinoxalin-2-one
C1=CC=C2C(=C1)NC(=O)C=N2
146.149
CHEBI:38890
Crystal-Powder at 20°C
2-Hydroxyquinoxaline
275°C
1196-57-2
MFCD00006722
FFRYUAVNPBUEIC-UHFFFAOYSA-N
1H-quinoxalin-2-one
14526
146.15
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6201

RTECSNumber : VD3630000

TSCA : Yes