Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxyquinoline, 99%

$51.23 - $51.23

Chemical Identifiers

Chemical Name or Material 2-Hydroxyquinoline
CAS 59-31-4
Molecular Formula C9H7NO
Formula Weight 145.16
MDL Number MFCD00006743
Beilstein 2855
Merck Index 14,1825
Synonym Carbostyril; 2-Quinolinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2306403

  SDS
alfa aesar™
B2306403
1g Each for $51.23
Description & Specifications

Specifications

Formula Weight 145.16
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Molecular Formula C9H7NO
CAS 59-31-4
Synonym Carbostyril; 2-Quinolinol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
MDL Number MFCD00006743
Merck Index 14,1825
Melting Point 196° to 199°C
TSCA TSCA
Beilstein 2855
Chemical Name or Material 2-Hydroxyquinoline
RTECS Number FG7175000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.