missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxyquinoline-4-carboxylic Acid, TCI America™

$161.95 - $485.84

Chemical Identifiers

CAS 15733-89-8
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00023942
InChI Key MFSHNFBQNVGXJX-UHFFFAOYSA-N
Synonym 2-hydroxyquinoline-4-carboxylic acid, 2-oxo-1,2-dihydroquinoline-4-carboxylic acid, 2-hydroxy-4-quinolincarboxylic acid, 2-hydroxycinchoninic acid, cinchoninic acid, 2-hydroxy, 4-carboxycarbostyril, 2-oxocinchoninic acid, 4-quinolinecarboxylic acid, 1,2-dihydro-2-oxo, 2-hydroxy-quinoline-4-carboxylic acid, 2-hydroxy-4-quinolinecarboxylic acid
PubChem CID 85076
ChEBI CHEBI:52045
IUPAC Name 2-oxo-1H-quinoline-4-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC(=O)N2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H10165G View Documents TCI America
H10165G
5g Each for $161.95
 
H101625G View Documents TCI America
H101625G
25g Each for $485.84
 
Specifications

Chemical Identifiers

15733-89-8
189.17
MFSHNFBQNVGXJX-UHFFFAOYSA-N
85076
2-oxo-1H-quinoline-4-carboxylic acid
C10H7NO3
MFCD00023942
2-hydroxyquinoline-4-carboxylic acid, 2-oxo-1,2-dihydroquinoline-4-carboxylic acid, 2-hydroxy-4-quinolincarboxylic acid, 2-hydroxycinchoninic acid, cinchoninic acid, 2-hydroxy, 4-carboxycarbostyril, 2-oxocinchoninic acid, 4-quinolinecarboxylic acid, 1,2-dihydro-2-oxo, 2-hydroxy-quinoline-4-carboxylic acid, 2-hydroxy-4-quinolinecarboxylic acid
CHEBI:52045
C1=CC=C2C(=C1)C(=CC(=O)N2)C(=O)O

Specifications

Yellow
15733-89-8
MFCD00023942
MFSHNFBQNVGXJX-UHFFFAOYSA-N
2-oxo-1H-quinoline-4-carboxylic acid
85076
189.17
≥98.0% (HPLC,T)
5g
C10H7NO3
2-hydroxyquinoline-4-carboxylic acid, 2-oxo-1,2-dihydroquinoline-4-carboxylic acid, 2-hydroxy-4-quinolincarboxylic acid, 2-hydroxycinchoninic acid, cinchoninic acid, 2-hydroxy, 4-carboxycarbostyril, 2-oxocinchoninic acid, 4-quinolinecarboxylic acid, 1,2-dihydro-2-oxo, 2-hydroxy-quinoline-4-carboxylic acid, 2-hydroxy-4-quinolinecarboxylic acid
C1=CC=C2C(=C1)C(=CC(=O)N2)C(=O)O
189.17
CHEBI:52045
Crystal-Powder at 20°C
2-Hydroxyquinoline-4-carboxylic Acid
Safety and Handling

Safety and Handling

RTECSNumber : GD3869000

TSCA : No